Merck
  • A Novel and Validated Protocol for Performing MIC Tests to Determine the Susceptibility of
A Novel and Validated Protocol for Performing MIC Tests to Determine the Susceptibility of
Frontiers in microbiology (2017-07-22)
Sergio Contreras-Lynch, Peter Smith, Paola Olmos, María E Loy, William Finnegan, Claudio D Miranda
ABSTRACT
This paper presents a validated protocol, using a novel, specifically formulated medium, to perform broth microdilution antimicrobial susceptibility assays of the salmonid bacterial pathogen
Product Description
Supelco
Florfenicol, analytical standard, for drug analysis