Merck
GABAergic mechanisms regulated by miR-33 encode state-dependent fear.
Nature neuroscience (2015-08-19)
Vladimir Jovasevic, Kevin A Corcoran, Katherine Leaderbrand, Naoki Yamawaki, Anita L Guedea, Helen J Chen, Gordon M G Shepherd, Jelena Radulovic
ABSTRACT
Fear-inducing memories can be state dependent, meaning that they can best be retrieved if the brain states at encoding and retrieval are similar. Restricted access to such memories can present a risk for psychiatric disorders and hamper their treatment. To better understand the mechanisms underlying state-dependent fear, we used a mouse model of contextual fear conditioning. We found that heightened activity of hippocampal extrasynaptic GABAA receptors, believed to impair fear and memory, actually enabled their state-dependent encoding and retrieval. This effect required protein kinase C-βII and was influenced by miR-33, a microRNA that regulates several GABA-related proteins. In the extended hippocampal circuit, extrasynaptic GABAA receptors promoted subcortical, but impaired cortical, activation during memory encoding of context fear. Moreover, suppression of retrosplenial cortical activity, which normally impairs retrieval, had an enhancing effect on the retrieval of state-dependent fear. These mechanisms can serve as treatment targets for managing access to state-dependent memories of stressful experiences.
Sigma-Aldrich
Anti-K+/Cl- Cotransporter (KCC2) Antibody, clone N1/12, clone N1/12, from mouse
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, ERK-2) antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Anti-CaM Kinase IIα (CaMKIIα) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-NR2A Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid