Merck
  • Astroglial connexin43 contributes to neuronal suffering in a mouse model of Alzheimer's disease.
Astroglial connexin43 contributes to neuronal suffering in a mouse model of Alzheimer's disease.
Cell death and differentiation (2016-07-09)
C Yi, X Mei, P Ezan, S Mato, I Matias, C Giaume, A Koulakoff
ABSTRACT
In Alzheimer's disease (AD), astrocyte properties are modified but their involvement in this pathology is only beginning to be appreciated. The expression of connexins, proteins forming gap junction channels and hemichannels, is increased in astrocytes contacting amyloid plaques in brains of AD patients and APP/PS1 mice. The consequences on their channel functions was investigated in a murine model of familial AD, the APPswe/PS1dE9 mice. Whereas gap junctional communication was not affected, we revealed that hemichannels were activated in astrocytes of acute hippocampal slices containing Aβ plaques. Such hemichannel activity was detected in all astrocytes, whatever their distance from amyloid plaques, but with an enhanced activity in the reactive astrocytes contacting amyloid plaques. Connexin43 was the main hemichannel contributor, however, a minor pannexin1 component was also identified in the subpopulation of reactive astrocytes in direct contact with plaques. Distinct regulatory pathways are involved in connexin and pannexin hemichannel activation. Inflammation triggered pannexin hemichannel activity, whereas connexin43 hemichannels were activated by the increase in resting calcium level of astrocytes. Importantly, hemichannel activation led to the release of ATP and glutamate that contributed to maintain a high calcium level in astrocytes placing them in the center of a vicious circle. The astroglial targeted connexin43 gene knocking-out in APPswe/PS1dE9 mice allowed to diminish gliotransmitter release and to alleviate neuronal damages, reducing oxidative stress and neuritic dystrophies in hippocampal neurons associated to plaques. Altogether, these data highlight the importance of astroglial hemichannels in AD and suggest that blocking astroglial hemichannel activity in astrocytes could represent an alternative therapeutic strategy in AD.
Sigma-Aldrich
10Panx1 trifluoroacetate salt, ≥98% (HPLC)
Sigma-Aldrich
Scrambled 10Panx1 trifluoroacetate salt, ≥98% (HPLC)