Merck
  • Molecular ion battery: a rechargeable system without using any elemental ions as a charge carrier.
Molecular ion battery: a rechargeable system without using any elemental ions as a charge carrier.
Scientific reports (2015-06-05)
Masaru Yao, Hikaru Sano, Hisanori Ando, Tetsu Kiyobayashi
ABSTRACT
Is it possible to exceed the lithium redox potential in electrochemical systems? It seems impossible to exceed the lithium potential because the redox potential of the elemental lithium is the lowest among all the elements, which contributes to the high voltage characteristics of the widely used lithium ion battery. However, it should be possible when we use a molecule-based ion which is not reduced even at the lithium potential in principle. Here we propose a new model system using a molecular electrolyte salt with polymer-based active materials in order to verify whether a molecular ion species serves as a charge carrier. Although the potential of the negative-electrode is not yet lower than that of lithium at present, this study reveals that a molecular ion can work as a charge carrier in a battery and the system is certainly a molecular ion-based "rocking chair" type battery.
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Tetrabutylammonium hexafluorophosphate, 98%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
1,5-Dibromopentane, 97%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Ammonium hexafluorophosphate, 99.98% trace metals basis
Sigma-Aldrich
Ammonium hexafluorophosphate, ≥95%
Sigma-Aldrich
4,4′−Dipyridyl, 98%
Sigma-Aldrich
Propylene carbonate, anhydrous, 99.7%