Merck
  • Excess SMAD signaling contributes to heart and muscle dysfunction in muscular dystrophy.
Excess SMAD signaling contributes to heart and muscle dysfunction in muscular dystrophy.
Human molecular genetics (2014-07-30)
Jeffery A Goldstein, Sasha Bogdanovich, Anastasia Beiriger, Lisa M Wren, Ann E Rossi, Quan Q Gao, Brandon B Gardner, Judy U Earley, Jeffery D Molkentin, Elizabeth M McNally
ABSTRACT
Disruption of the dystrophin complex causes muscle injury, dysfunction, cell death and fibrosis. Excess transforming growth factor (TGF) β signaling has been described in human muscular dystrophy and animal models, where it is thought to relate to the progressive fibrosis that characterizes dystrophic muscle. We now found that canonical TGFβ signaling acutely increases when dystrophic muscle is stimulated to contract. Muscle lacking the dystrophin-associated protein γ-sarcoglycan (Sgcg null) was subjected to a lengthening protocol to produce maximal muscle injury, which produced rapid accumulation of nuclear phosphorylated SMAD2/3. To test whether reducing SMAD signaling improves muscular dystrophy in mice, we introduced a heterozygous mutation of SMAD4 (S4) into Sgcg mice to reduce but not ablate SMAD4. Sgcg/S4 mice had improved body mass compared with Sgcg mice, which normally show a wasting phenotype similar to human muscular dystrophy patients. Sgcg/S4 mice had improved cardiac function as well as improved twitch and tetanic force in skeletal muscle. Functional enhancement in Sgcg/S4 muscle occurred without a reduction in fibrosis, suggesting that intracellular SMAD4 targets may be important. An assessment of genes differentially expressed in Sgcg muscle focused on those encoding calcium-handling proteins and responsive to TGFβ since this pathway is a target for mediating improvement in muscular dystrophy. These data demonstrate that excessive TGFβ signaling alters cardiac and muscle performance through the intracellular SMAD pathway.
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium chloride, tablet