Merck
  • Expression of a novel stress-inducible protein, sestrin 2, in rat glomerular parietal epithelial cells.
Expression of a novel stress-inducible protein, sestrin 2, in rat glomerular parietal epithelial cells.
American journal of physiology. Renal physiology (2014-07-25)
Hiroko Hamatani, Keiju Hiromura, Toru Sakairi, Satoshi Takahashi, Mitsuharu Watanabe, Akito Maeshima, Takamoto Ohse, Jeffery W Pippin, Stuart J Shankland, Yoshihisa Nojima
ABSTRACT
Sestrin 2, initially identified as a p53 target protein, accumulates in cells exposed to stress and inhibits mammalian target of rapamycin (mTOR) signaling. In normal rat kidneys, sestrin 2 was selectively expressed in parietal epithelial cells (PECs), identified by the marker protein gene product 9.5. In adriamycin nephropathy, sestrin 2 expression decreased in PECs on day 14, together with increased expression of phosphorylated S6 ribosomal protein (P-S6RP), a downstream target of mTOR. Sestrin 2 expression was markedly decreased on day 42, coinciding with glomerulosclerosis and severe periglomerular fibrosis. In puromycin aminonucleoside nephropathy, decreased sestrin 2 expression, increased P-S6RP expression, and periglomerular fibrosis were observed on day 9, when massive proteinuria developed. These changes were transient and nearly normalized by day 28. In crescentic glomerulonephritis, sestrin 2 expression was not detected in cellular crescents, whereas P-S6RP increased. In conditionally immortalized cultured PECs, the forced downregulation of sestrin 2 by short hairpin RNA resulted in increased expression of P-S6RP and increased apoptosis. These data suggest that sestrin 2 is involved in PEC homeostasis by regulating the activity of mTOR. In addition, sestrin 2 could be a novel marker of PECs, and decreased expression of sestrin 2 might be a marker of PEC injury.
Product Description
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Periodic acid, 99.999% trace metals basis
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
Periodic acid, SAJ special grade, ≥98.5%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sesn2
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Periodic acid, suitable for electrophoresis, ≥99%
SAFC
Sodium deoxycholate
Sigma-Aldrich
Periodic acid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Periodic acid, puriss. p.a., ACS reagent, for oxidimetric titration, crystallized, ≥99.0% (RT)
Sigma-Aldrich
Periodic acid, ACS reagent, 99%