Merck
  • Prognostic significance of cyclooxygenase-2 pathway and angiogenesis in head and neck squamous cell carcinoma.
Prognostic significance of cyclooxygenase-2 pathway and angiogenesis in head and neck squamous cell carcinoma.
Human pathology (2002-08-28)
Oreste Gallo, Emanuela Masini, Benedetta Bianchi, Luca Bruschini, Milena Paglierani, Alessandro Franchi
ABSTRACT
Prostaglandins play a critical role in tumor development and growth by regulating numerous biologic processes, including tumor angiogenesis, with clear prognostic and therapeutic implications. The aim of this study was to investigate the prognostic relevance of cyclooxygenase-2 (COX-2) pathway activation in head and neck squamous cell carcinoma (HNSCC). COX-2 activity was analyzed in 52 consecutive patients by assessing protein expression and prostaglandin E(2) (PgE(2)) levels and was then correlated to vascular endothelial growth factor (VEGF) expression and tumor angiogenesis. We evaluated the prognostic impact of these parameters by Kaplan-Meier and Cox survival analysis. COX-2 expression by tumor cells was closely correlated to VEGF expression and to tumor vascularization. According to Kaplan-Meier analysis, patients with COX-2 tumor overexpression and with higher PgE(2) tumor levels had significantly shorter overall survival estimates (P = 0.022 and P = 0.033, respectively). Analogously, patients with more-vascularized tumors had worse survival than those with less-vascularized cancers (P = 0.032). Cox multivariate analysis demonstrated that the most significant prognostic factors were presence of lymph node metastasis, tumor vascularization, COX-2 protein expression, and PgE(2) tumor levels. This study demonstrates a close correlation between COX-2 pathway, VEGF expression, and tumor angiogenesis in HNSCC. In addition, COX-2 overexpression and higher tumor vascularization appear to predict a shorter survival in patients with head and neck cancer.
Sigma-Aldrich
COX-2 (SP21) Rabbit Monoclonal Antibody