Merck
  • A novel PNA-monomer for recognition of thymine in triple-helix structures.
A novel PNA-monomer for recognition of thymine in triple-helix structures.
Nucleosides, nucleotides & nucleic acids (2003-10-21)
Anne Goldbech Olsen, Otto Dahl, Peter E Nielsen
ABSTRACT
To expand the triplex recognition repertoire of Nucleic Acids, novel nucleobases that recognize thymine in a T-A base pair are still required. A novel conformationally constrained PNA-monomer (II) capable of binding T in a triplex motif was designed and synthesized in 7 steps starting from commercially available dimethyl 2-oxoglutarate.
Sigma-Aldrich
Dimethyl 2-oxoglutarate, 96%