Merck
Side-firing Neodymium:YAG laser prostatectomy.
European urology (1999-02-06)
A J Costello, J N Kabalin
ABSTRACT
Not until 1991 did laser therapy for benign prostatic hyperplasia become a feasible option for the practicing urologist. Prior to the development of side firing free beam laser delivery devices contact Neodymium:YAG, laser therapy for BPH was of limited applicability. Since 1991, refinements in laser device technology have allowed the urologist to expand the potential for Neodymium:YAG coagulative prostatectomy. Multiple studies over the past five years have demonstrated equivalence in efficacy and safety to conventional electrocautery TURP. Holmium:YAG laser technology may be a significant advance again on free beam side firing Neodymium:YAG laser prostatectomy.
Product Description
Neodymium, rod, 100mm, diameter 12.5mm, cast, 99%
Neodymium, rod, 100mm, diameter 6.35mm, cast, 99%
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis
Neodymium, rod, 50mm, diameter 12.5mm, cast, 99%
Neodymium, rod, 50mm, diameter 6.35mm, cast, 99%