Merck
  • Applications of the neodymium:YAG laser in plastic surgery of the face and lacrimal surgery. Wound repair. A review.
Applications of the neodymium:YAG laser in plastic surgery of the face and lacrimal surgery. Wound repair. A review.
Ophthalmologica. Journal international d'ophtalmologie. International journal of ophthalmology. Zeitschrift fur Augenheilkunde (2003-02-05)
Franz Fankhauser, Sylwia Kwasniewska
ABSTRACT
The physical and clinical features of a number of laser energy sources presently being used in plastic and endonasal surgery, such as the Nd:YAG, KTP, Ho:YAG, Er:YAG and CO(2) lasers, are emphasized. Particular attention is paid to the Nd:YAG laser and to an Nd:YAG laser-powered quartz laser scalpel.
Product Description
Neodymium, rod, 50mm, diameter 12.5mm, cast, 99%
Neodymium, rod, 50mm, diameter 6.35mm, cast, 99%
Neodymium, rod, 100mm, diameter 12.5mm, cast, 99%
Neodymium, rod, 100mm, diameter 6.35mm, cast, 99%
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis