Merck
  • A structure-activity relationship study of 1,2,4-triazolo[1,5-a][1,3,5]triazin-5,7-dione and its 5-thioxo analogues on anti-thymidine phosphorylase and associated anti-angiogenic activities.
A structure-activity relationship study of 1,2,4-triazolo[1,5-a][1,3,5]triazin-5,7-dione and its 5-thioxo analogues on anti-thymidine phosphorylase and associated anti-angiogenic activities.
European journal of medicinal chemistry (2013-07-23)
Hriday Bera, Bee Jen Tan, Lingyi Sun, Anton V Dolzhenko, Wai-Keung Chui, Gigi Nagar Chee Chiu
ABSTRACT
Thirty-three 1,2,4-triazolo[1,5-a][1,3,5]triazin-5,7-dione and its 5-thioxo analogues were designed and synthesized which contained different substituents at meta- and/or para-positions of 2-phenyl or 2-benzyl ring attached to the fused ring structure. The preliminary pharmacological evaluation demonstrated that the 5-thioxo analogues of 1,2,4-triazolo[1,5-a][1,3,5]triazine exhibited a varying degree of inhibitory activity towards thymidine phosphorylase, comparable or better than reference compound, 7-Deazaxanthine (7-DX, 2) (IC50 value = 42.63 μM). Moreover, compounds 5q and 6i displayed a mixed-type of inhibitory mechanism in the presence of variable concentrations of thymidine (dThd). In addition, selected compounds were found to have a noticeable inhibitory effect on the expression of angiogenesis markers, including VEGF and MMP-9 in MDA-MB-231 breast cancer cells.
Sigma-Aldrich
Thymidine Phosphorylase, recombinant from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution, ≥500 units/mL
Sigma-Aldrich
Thymidine Phosphorylase, recombinant from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution, ≥900 units/mL, 0.2 μm filtered
Sigma-Aldrich
Thymidine Phosphorylase, recombinant from Escherichia coli, recombinant, expressed in E. coli, Suitable for manufacturing of diagnostic kits and reagents, buffered aqueous solution, ≥500 units/mL