Merck
  • Heparin-bonded covered stents versus bare-metal stents for complex femoropopliteal artery lesions: the randomized VIASTAR trial (Viabahn endoprosthesis with PROPATEN bioactive surface [VIA] versus bare nitinol stent in the treatment of long lesions in superficial femoral artery occlusive disease).
Heparin-bonded covered stents versus bare-metal stents for complex femoropopliteal artery lesions: the randomized VIASTAR trial (Viabahn endoprosthesis with PROPATEN bioactive surface [VIA] versus bare nitinol stent in the treatment of long lesions in superficial femoral artery occlusive disease).
Journal of the American College of Cardiology (2013-07-09)
Johannes Lammer, Thomas Zeller, Klaus A Hausegger, Philipp J Schaefer, Manfred Gschwendtner, Stefan Mueller-Huelsbeck, Thomas Rand, Martin Funovics, Florian Wolf, Aljoscha Rastan, Michael Gschwandtner, Stefan Puchner, Robin Ristl, Maria Schoder
ABSTRACT
The hypothesis that endovascular treatment with covered stents has equal risks but higher efficacy than bare-metal stents (BMS) in long femoropopliteal artery disease was tested. Although endovascular treatment of short superficial femoral artery lesions revealed excellent results, efficacy in long lesions remains unsatisfactory. In a prospective, randomized, single-blind, multicenter study, 141 patients with symptomatic peripheral arterial disease were assigned to treatment with heparin-bonded, covered stents (Viabahn 72 patients) or BMS (69 patients). Clinical outcomes and patency rates were assessed at 1, 6, and 12 months. Mean ± SD lesion length was 19.0 ± 6.3 cm in the Viabahn group and 17.3 ± 6.6 cm in the BMS group. Major complications within 30 days were observed in 1.4%. The 12-month primary patency rates in the Viabahn and BMS groups were: intention-to-treat (ITT) 70.9% (95% confidence interval [CI]: 0.58 to 0.80) and 55.1% (95% CI: 0.41 to 0.67) (log-rank test p = 0.11); treatment per-protocol (TPP) 78.1% (95% CI: 0.65 to 0.86) and 53.5% (95% CI: 0.39 to 0.65) (hazard ratio: 2.23 [95% CI: 1.14 to 4.34) (log-rank test p = 0.009). In lesions ≥20 cm, (TransAtlantic Inter-Society Consensus class D), the 12-month patency rate was significantly longer in VIA patients in the ITT analysis (VIA 71.3% vs. BMS 36.8%; p = 0.01) and the TPP analysis (VIA 73.3% vs. BMS 33.3%; p = 0.004). Freedom from target lesion revascularization was 84.6% for Viabahn (95% CI: 0.72 to 0.91) versus 77.0% for BMS (95% CI: 0.63 to 0.85; p = 0.37). The ankle-brachial index in the Viabahn group significantly increased to 0.94 ± 0.23 compared with the BMS group (0.85 ± 0.23; p < 0.05) at 12 months. This randomized trial in symptomatic patients with peripheral arterial disease who underwent endovascular treatment for long femoropopliteal lesions demonstrated significant clinical and patency benefits for heparin-bonded covered stents compared with BMS in lesions ≥20 cm and for all lesions in the TPP analysis. In the ITT analysis for all lesions, which was flawed by major protocol deviations in 8.5% of the patients, the difference was not significant. (GORE VIABAHN® endoprosthesis with bioactive propaten surface versus bare nitinol stent in the treatment of TASC B, C and D lesions in superficial femoral artery occlusive disease; ISRCTN48164244).
Sigma-Aldrich
Polytetrafluoroethylene preparation, 60 wt % dispersion in H2O
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size