Merck
  • IL-33 promotes airway remodeling in pediatric patients with severe steroid-resistant asthma.
IL-33 promotes airway remodeling in pediatric patients with severe steroid-resistant asthma.
The Journal of allergy and clinical immunology (2013-06-14)
Sejal Saglani, Stephen Lui, Nicola Ullmann, Gaynor A Campbell, Rebekah T Sherburn, Sara A Mathie, Laura Denney, Cara J Bossley, Timothy Oates, Simone A Walker, Andrew Bush, Clare M Lloyd
ABSTRACT
TH2 cytokines are not responsible for the ongoing symptoms and pathology in children with severe therapy-resistant asthma (STRA). IL-33 induces airway hyperresponsiveness, but its role in airway remodeling and steroid resistance is unknown. We sought to investigate the relationship between IL-33 and airway remodeling in pediatric patients with STRA. IL-33 levels were quantified in neonatal mice given inhaled house dust mite (HDM), and the effect of blocking IL-13 on remodeling and IL-33 levels was assessed. HDM-induced allergic airways disease (AAD) in neonatal ST2(-/-) mice lacking the IL-33 receptor was assessed, together with collagen production after IL-33 administration. The effect of steroid therapy on IL-33 levels in patients with neonatal AAD was explored. IL-33 expression was quantified in endobronchial biopsy (EB) specimens from children with STRA and related to remodeling, and collagen production by airway fibroblasts from pediatric patients stimulated with IL-33 and budesonide was quantified. Blocking IL-13 after AAD was established in neonatal mice and did not reduce remodeling or IL-33 levels; airway hyperresponsiveness was only partially reduced. IL-33 promoted collagen synthesis both from asthmatic fibroblasts from pediatric patients and after intranasal administration in mice. Increased cellular expression of IL-33, but not IL-13, was associated with increased reticular basement membrane thickness in EB specimens from children with STRA, whereas remodeling was absent in HDM-exposed ST2(-/-) mice. IL-33 levels were maintained, whereas IL-13 levels were abrogated by steroid treatment in neonatal HDM-exposed mice and in EB specimens from children with STRA. IL-33 is a relatively steroid-resistant mediator that promotes airway remodeling in patients with STRA and is an important therapeutic target.
Sigma-Aldrich
Budesonide, ≥99%
Budesonide, European Pharmacopoeia (EP) Reference Standard
Supelco
Budesonide, Pharmaceutical Secondary Standard; Certified Reference Material