Merck
  • Poly(C)-binding protein 1, a novel N(pro)-interacting protein involved in classical swine fever virus growth.
Poly(C)-binding protein 1, a novel N(pro)-interacting protein involved in classical swine fever virus growth.
Journal of virology (2012-12-12)
Dan Li, Su Li, Yuan Sun, Hong Dong, Yongfeng Li, Bibo Zhao, Dongwei Guo, Changjiang Weng, Hua-Ji Qiu
ABSTRACT
N(pro) is a multifunctional autoprotease unique to pestiviruses. The interacting partners of the N(pro) protein of classical swine fever virus (CSFV), a swine pestivirus, have been insufficiently defined. Using a yeast two-hybrid screen, we identified poly(C)-binding protein 1 (PCBP1) as a novel interacting partner of the CSFV N(pro) protein and confirmed this by coimmunoprecipitation, glutathione S-transferase (GST) pulldown, and confocal assays. Knockdown of PCBP1 by small interfering RNA suppressed CSFV growth, while overexpression of PCBP1 promoted CSFV growth. Furthermore, we showed that type I interferon was downregulated by PCBP1, as well as N(pro). Our results suggest that cellular PCBP1 positively modulates CSFV growth.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M1 antibody, clone M1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution