Merck
  • [Comparison of anti-inflammatory effect and analysis of astilbin red and white transverse section Smilax glabra in 28 collection sites].
[Comparison of anti-inflammatory effect and analysis of astilbin red and white transverse section Smilax glabra in 28 collection sites].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-03-13)
Xi-Cheng He, Qin-Wen Shun, Li-Sha Dong, Xiang-Qian Ge, Hua Zhang, Heng Nong, Xiao-Hua Shen
ABSTRACT
Astilbin in 28 Smilax glabra (red and white cross-section) from different sources was determined by HPLC. Pharmacodynamics and component of S. glabra was investigated through inflammation experiment (penetration type). The analysis was performed on a Hypersil ODS2 column (4.6 mm x 250 mm, 5 microm) with the mobile phase of acetonitrile and 0. 1% acetic acid aqueous (21: 79) at a flow rate of 1.0 mL x min(-1). The detection wavelength was 291 nm, and the column temperature was 25 degrees C. Anti-inflammatory effect was compared from two type cross-section of Smilax glabra in capillary permeability experiment. Linear correlation was good in the range of 0.003 379-4.004 microg, and the average recoveries were 100.1%, 101.9%, 99.3%, respectively. The content of astilbin in white and red transverse section were 0.19%-2.46% and 2.10%-5.92%, respectively. Anti-inflammatory efficiency of sectioned red and white were were 21% and 32%, respectively. Astilbin content and anti-inflammatory effect is significantly different between red and white transverse section of S. glabra, the content of astilbin is not positively related with anti-inflammatory effect.
Sigma-Aldrich
Astilbin from Engelhardtia roxburghiana, ≥98%