Merck
  • Enantioselective synthesis of (+)-isolysergol via ring-closing metathesis.
Enantioselective synthesis of (+)-isolysergol via ring-closing metathesis.
Organic letters (2010-05-14)
Jason A Deck, Stephen F Martin
ABSTRACT
The first enantioselective synthesis of (+)-isolysergol was completed in 12 steps from commercially available materials by a novel approach that features a late stage microwave-mediated, diastereomeric ring-closing metathesis catalyzed by a chiral molybdenum catalyst to simultaneously form the D ring and set the stereocenter at C(8).
Sigma-Aldrich
1,4-Pentadiene, 99%