Merck
  • The pH-dependent leaching of inorganic contaminants from secondary lead smelter fly ash.
The pH-dependent leaching of inorganic contaminants from secondary lead smelter fly ash.
Journal of hazardous materials (2009-02-07)
Martina Vítková, Vojtech Ettler, Ondrej Sebek, Martin Mihaljevic, Tomás Grygar, Jan Rohovec
ABSTRACT
The leaching behaviour of fly ash (FA) from a secondary Pb smelter was assessed using the pH-static leaching experiment according to prEN 14997 (pH range 3-11) coupled with mineralogical investigation of the leached FA by XRD and Rietveld analyses and thermodynamic modelling using PHREEQC-2. The procedure was performed on fresh FA and FA washed at a cumulative L/S ratio of 60l/kg to remove readily soluble salts. For both fresh and washed FA, high amounts of inorganic contaminants were released under acidic conditions, exhibiting L-shaped leaching patterns: up to 300g Pb/kg, 4.5g Cd/kg, 4g Zn/kg, 1.05g As/kg and 70mg Sb/kg. The washing of soluble salts significantly decreased the leachability of Cd, Zn, As and Sb and increased the release of Pb, especially under acidic conditions. The leaching of fresh FA removed part of primary caracolite and all the KPb(2)Cl(5) and NaCl. The Pb release was controlled by the precipitation of anglesite and PbSO(3) under acidic conditions and of laurionite and carbonates (hydrocerussite and phosgenite) under alkaline conditions. In contrast, the washed FA was composed mainly of anglesite and PbSO(3), both phases being the main solubility-controlling phases for Pb over the whole studied pH range.
Sigma-Aldrich
Lead(II) sulfate, 98%
Sigma-Aldrich
Lead(II) sulfate, 99.99% trace metals basis