Merck
  • Conformational analysis of dynorphin A (1-13) using hydrogen-deuterium exchange and tandem mass spectrometry.
Conformational analysis of dynorphin A (1-13) using hydrogen-deuterium exchange and tandem mass spectrometry.
European journal of mass spectrometry (Chichester, England) (2008-04-18)
Xianmei Cai, Chhabil Dass
ABSTRACT
Trifluoroethanol (TFE)-induced conformational changes in dynorphin A (1-13) were investigated using charge-state distribution (CSD) and hydrogen-deuterium exchange (HDX), combined with electrospray ionization (ESI) mass spectrometry (MS). Individual amino acids involved in secondary structural elements were identified by collision-induced dissociation-tandem mass spectrometry (MS/MS). It was observed that dynorphin A (1-13) largely exists in an unfolded conformation and a folded structure in increasing concentrations of TFE. In 50% TFE, it forms an alpha-helix that encompasses residues 1-9 and remains flexible from residues 10 to 13.
Sigma-Aldrich
Dynorphin A Porcine Fragment 1-13, ≥97% (HPLC)