Merck
Caffeine gum minimizes sleep inertia.
Perceptual and motor skills (2013-07-09)
Rachel A Newman, Gary H Kamimori, Nancy J Wesensten, Dante Picchioni, Thomas J Balkin
ABSTRACT
Naps are an effective strategy for maintaining alertness and cognitive performance; however, upon abrupt wakening from naps, sleep inertia (temporary performance degradation) may ensue. In the present study, attenuation of post-nap sleep inertia was attempted by administration of caffeine gum. Using a double-blind, placebo-controlled crossover design, 15 healthy, non-smoking adults were awakened at 1 hr. and again at 6 hr. after lights out (0100 and 0600, respectively) and were immediately administered a gum pellet containing 100 mg of caffeine or placebo. A 5-min. psychomotor vigilance task was administered at 0 min., 6 min., 12 min., and 18 min. post-awakening. At 0100, response speed with caffeine was significantly better at 12 min. and 18 min. post-awakening compared to placebo; at 0600, caffeine's effects were evident at 18 min. post-awakening. Caffeinated gum is a viable means of rapidly attenuating sleep inertia, suggesting that the adenosine receptor system is involved in sleep maintenance.
Product Description
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, BioXtra