Merck
Effective management of exudate with AQUACEL extra.
British journal of community nursing (2013-05-03)
Joy Tickle
ABSTRACT
The management of patients with highly-exuding wounds can often lead to the use of unreliable and costly treatments. Patients are frequently found to be at increased risk of infection and delayed healing, which results in a huge negative impact on their quality of life. In the political and health arena, reducing health-care costs yet maintaining high quality and productivity is high on the list. With current economic, health-care and political targets, it is crucial to address the importance of effective wound management, particularly the importance of managing exudate. Inappropriate management of wound exudate can lead to prolonged wound healing, peri-wound maceration and excoriation, and can result in deterioration of the wound.
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~700,000
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Carboxymethylcellulose sodium salt, SAJ first grade
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity