Merck
  • Down-regulation of signal transducer and activator of transcription 3 improves human acute myeloid leukemia-derived dendritic cell function.
Down-regulation of signal transducer and activator of transcription 3 improves human acute myeloid leukemia-derived dendritic cell function.
Leukemia research (2013-05-01)
Michael T Brady, Austin Miller, Sheila N Sait, Laurie A Ford, Hans Minderman, Eunice S Wang, Kelvin P Lee, Heinz Baumann, Meir Wetzler
ABSTRACT
Signal transducer and activator of transcription (STAT) 3 inhibits dendritic cell (DC) differentiation and is constitutively activated in blasts of approximately half of AML patients. We investigated the correlation between STAT3 activity, DC maturation and the ability to stimulate T-cells in primary acute myeloid leukemia (AML)-derived DCs. STAT3 knock-down by shRNAmir increased the ability of AML-DCs to stimulate T-cells. Treatment of AML-DC with arsenic trioxide, but not AG490, JSI-124 or NSC-74859, led to a more mature phenotype and enhanced T-cell stimulation, while having minimal effect on normal DC. We conclude that AML-DCs have improved immunogenicity after reducing STAT3.
Sigma-Aldrich
Arsenic(III) oxide, SAJ first grade, ≥99.0%
Supelco
Arsenic(III) oxide, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Arsenic(III) oxide, 99.995% trace metals basis
Sigma-Aldrich
Arsenic(III) oxide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Arsenic(III) oxide, ACS reagent (primary standard)