Merck
  • High titer ethanol production from SPORL-pretreated lodgepole pine by simultaneous enzymatic saccharification and combined fermentation.
High titer ethanol production from SPORL-pretreated lodgepole pine by simultaneous enzymatic saccharification and combined fermentation.
Bioresource technology (2012-11-10)
T Q Lan, Roland Gleisner, J Y Zhu, Bruce S Dien, Ronald E Hector
ABSTRACT
Lodgepole wood chips were pretreated by sulfite pretreatment to overcome recalcitrance of lignocelluloses (SPORL) at 25% solids loading and 180 °C for 20 min with sulfuric acid and sodium bisulfite charges of 2.2 and 8 wt/wt% on an oven-dry wood basis, respectively. The pretreated wood chips were disk-milled with pretreatment spent liquor and water, and the solid fraction was separated from the liquor stream. The liquor was neutralized and concentrated through vacuum evaporation. Quasi-simultaneous enzymatic saccharification of the cellulosic solids and combined fermentation with the concentrated liquor was conducted at up to 20% total solids loading. Fed-batching of the solids facilitated liquefaction and saccharification, as well as managing instantaneous inhibitor concentrations. At a commercial cellulase (CTec2) loading of only 9 FPU or 0.06 mL/g untreated wood, a maximum ethanol titer of 47.4 g/L was achieved, resulting in a calculated yield of 285 L/tonne of wood using Saccharomyces cerevisiae YRH400 at 35 °C and pH 5.5.
Sigma-Aldrich
Sodium bisulfite, JIS special grade, 64.0-67.4% SO2 basis
Sigma-Aldrich
Sodium bisulfite, anhydrous, free-flowing, Redi-Dri, ACS reagent
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
Sigma-Aldrich
Sodium bisulfite, ACS reagent