Merck
  • Lesion of olfactory epithelium attenuates expression of morphine-induced behavioral sensitization and reinstatement of drug-primed conditioned place preference in mice.
Lesion of olfactory epithelium attenuates expression of morphine-induced behavioral sensitization and reinstatement of drug-primed conditioned place preference in mice.
Pharmacology, biochemistry, and behavior (2012-10-30)
Haichen Niu, Yingwei Zheng, Tanzeel Huma, Joshua D Rizak, Ling Li, Guimei Wang, He Ren, Liqi Xu, Jianzhen Yang, Yuanye Ma, Hao Lei
ABSTRACT
Previous studies have shown that olfactory impairment by disrupting the olfactory epithelium prior to morphine administration attenuated the development addiction-related behaviors. However, it is unclear whether olfactory impairment will affect the expression of already established addiction-related behaviors. To address this issue, mice were conditioned with morphine to induce behavioral sensitization and condition placed preference (CPP). After an abstinence period, the animals were subjected to either an intranasal ZnSO(4) effusion (ZnE) or sham treatment with saline. Behavioral sensitization and CPP reinstatement were evaluated 24h later, as well as the expression of c-Fos protein, a marker of activated neural sites, in brain regions of interest. It was found that ZnE treatment attenuated morphine-induced behavioral sensitization and reinstatement of CPP. Compared to the saline-treated ones, the ZnE-treated animals showed reduced c-Fos expression in the nucleus accumbens (NAc) associated with behavioral sensitization, and in the NAc, cingulate cortex, dentate gyrus, amygdala, lateral hypothalamus and ventral tegmental area associated with CPP reinstatement. Together, these results demonstrated that acute olfactory impairment could attenuate already established addiction-related behaviors and expression of c-Fos in drug addiction related brain regions, perhaps by affecting the coordination between reward and motivational systems in the brain.
Sigma-Aldrich
Zinc sulfate heptahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Zinc sulfate heptahydrate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Zinc sulfate heptahydrate, meets USP testing specifications
Sigma-Aldrich
Zinc sulfate heptahydrate, suitable for plant cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, ACS reagent, 99%
Sigma-Aldrich
Zinc sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Zinc sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Zinc sulfate heptahydrate, BioUltra, for molecular biology, 2.0 M in H2O
Sigma-Aldrich
Zinc sulfate solution, 0.3 N