Merck
  • Antidiarrhoeal efficacy of Mangifera indica seed kernel on Swiss albino mice.
Antidiarrhoeal efficacy of Mangifera indica seed kernel on Swiss albino mice.
Asian Pacific journal of tropical medicine (2012-07-31)
S Rajan, H Suganya, T Thirunalasundari, S Jeeva
ABSTRACT
To examine the antidiarrhoeal activity of alcoholic and aqueous seed kernel extract of Mangifera indica (M. indica) on castor oil-induced diarrhoeal activity in Swiss albino mice. Mango seed kernels were processed and extracted using alcohol and water. Antidiarrhoeal activity of the extracts were assessed using intestinal motility and faecal score methods. Aqueous and alcoholic extracts of M. indica significantly reduced intestinal motility and faecal score in Swiss albino mice. The present study shows the traditional claim on the use of M. indica seed kernel for treating diarrhoea in Southern parts of India.
Sigma-Aldrich
Castor oil, CP
Sigma-Aldrich
Castor oil
Sigma-Aldrich
Castor oil, meets testing specifications per USP
Sigma-Aldrich
Castor oil, tested according to Ph. Eur.