Merck
Two new compounds isolated from Liriope muscari.
Molecules (Basel, Switzerland) (2012-07-27)
Wen-Jie Li, Zhi-Hao Zhang, Xian-Long Cheng, Jing Liu, Yi He, Chao Zhou, Ying Guo, Rui-Chao Lin, Gang-Li Wang
ABSTRACT
Two new compounds, (2S,3R)-methyl 7-hydroxy-2-(4-hydroxy-3-methoxy-phenyl)-3-(hydroxymethyl)-2,3-dihydrobenzofuran-5-carboxylate (1) and (4R,5S)-5-(3-hydroxy-2,6-dimethylphenyl)-4-isopropyldihydrofuran-2-one (2), tentatively named norcurlignan and limlactone, respectively, were isolated from Liriope muscari, together with the known compound (-)-pinoresinol (3). The structures of these compounds were elucidated and characterized on the basis of 1D NMR, 2D NMR, CD and MS data. The in vitro antioxidant activities of compounds 1-3 were assessed by the DPPH and ABTS scavenging methods.
Product Description
Supelco
Pinoresinol, analytical standard
Sigma-Aldrich
Pinoresinol, ≥95.0% (HPLC)