Merck
  • Metal-free synthesis of 3,3-disubstituted oxoindoles by iodine(III)-catalyzed bromocarbocyclizations.
Metal-free synthesis of 3,3-disubstituted oxoindoles by iodine(III)-catalyzed bromocarbocyclizations.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-07-13)
David C Fabry, Maciej Stodulski, Stefanie Hoerner, Tanja Gulder
ABSTRACT
"I" did it: An iodine(III)-mediated bromocarbocyclization was elaborated as an efficient tool for the synthesis of oxoindoles. This method is applicable to a variety of structurally different substrates, also with chemically sensitive groups, and gives access to the heterocycles in a regio- and stereoselective fashion. The indole-2-ones obtained can be converted easily into structurally complex target compounds, such as the alkaloid physostigmine.
Sigma-Aldrich
Potassium bromide, JIS special grade, 99.0-100.2%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Methacrylamide, 98%