Merck
  • Cyclooxygenase-2 inhibition and thromboxane A(2) receptor antagonism attenuate hypoxic pulmonary vasoconstriction in a porcine model.
Cyclooxygenase-2 inhibition and thromboxane A(2) receptor antagonism attenuate hypoxic pulmonary vasoconstriction in a porcine model.
Acta physiologica (Oxford, England) (2012-05-05)
D Kylhammar, G Rådegran
ABSTRACT
Hypoxic pulmonary vasoconstriction (HPV) causes pulmonary hypertension that may lead to right heart failure. We hypothesized that the COX-2 inhibitor nimesulide and the thromboxane A(2) receptor antagonist daltroban would attenuate HPV. Haemodynamic measurements and blood sampling were performed in 18 anaesthetized, mechanically ventilated pigs, with mean ± SEM weights of 31.3 ± 0.6 kg, in normoxia (F(i)O(2)~0.21) and hypoxia (F(i)O(2)~0.10), before and 5, 15 and 45 min after initiation of right atrial infusion of nimesulide (n = 6) or daltroban (n = 6), respectively, and in six control pigs. Compared with normoxia, hypoxia (n = 18) increased mean pulmonary artery pressure by 15.8 ± 0.8 mmHg (P < 0.001), pulmonary vascular resistance (PVR) by 2.7 ± 0.3 WU (P < 0.05) and mean right atrial pressure by 2.3 ± 0.3 mmHg (P < 0.001). In the control pigs, mean pulmonary artery pressure, PVR and mean right atrial pressure remained stable (P = ns) throughout 45 min hypoxia, compared with hypoxia baseline. Nimesulide decreased mean pulmonary artery pressure by 3.7 ± 1.3 mmHg after 45 min (P < 0.013), as well as PVR by 0.8 ± 0.2 WU (P < 0.05), levelling off after 15 min. Daltroban transiently increased (P < 0.001) mean pulmonary artery pressure and mean right atrial pressure by 7.2 ± 1.2 and 2.7 ± 0.4 mmHg, respectively, but they returned to hypoxia baseline (P = ns) within 5 min. Daltroban then decreased mean pulmonary artery pressure to after 45 min be 4.2 ± 1.6 mmHg lower (P < 0.005) than at hypoxia baseline. COX-2 inhibition and thromboxane A(2) receptor antagonism attenuate HPV by decreasing mean pulmonary artery pressure by approximately 10-11%, as measured 45 min after initiation of nimesulide or daltroban infusion respectively.
Sigma-Aldrich
Nimesulide
Nimesulide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Daltroban, >98% (HPLC)