Merck
  • Novel two-component gels of cetylpyridinium chloride and the bola-amphiphile 6-amino caproic acid: phase evolution and mechanism of gel formation.
Novel two-component gels of cetylpyridinium chloride and the bola-amphiphile 6-amino caproic acid: phase evolution and mechanism of gel formation.
The journal of physical chemistry. B (2012-02-09)
Illa Ramakanth, Archita Patnaik
ABSTRACT
A two-component gel resulting from the amphiphilic cationic surfactant cetylpyridinium chloride (CPC) in the presence of a structure-forming bola-amphiphilic additive 6-aminocaproic acid (6-ACA) was realized and investigated. At a critical 6 wt % of 1:1 CPC:6-ACA, the yellow colored gel resulted from a 3:1 v/v CHCl(3):H(2)O critical binary solvent composition. The mixed amphiphilic system formed a 1:1 complex with a binding constant ~0.83 × 10(4) M(-1). Phase evolution and mechanism of gelation in the mixed CPC:6-ACA system was unraveled upon investigating the gel microstructure, based on spectroscopic, microscopic, and small-angle X-ray scattering (SAXS) investigations. The gel assembled as a lamellar organization, maintaining a loosely interdigitated bilayer structure of CPC and 6-ACA molecules through predominant charge transfer, H-bonding, and hydrophobic and intercomplex interactions. The SAXS pattern indicated a semicrystalline form to be the stable phase with alternating crystalline and amorphous layers; a novel mode of gelation with a widely disparate semicrystalline form of the lamellar gel was thus indicated, where the lamellar structure was deduced from the interplanar spacings. A transition from low viscosity reverse micellar solution to a yellow rigid gel upon aging was thus comprehended. The mixed amphiphile in varying polarity organic solvents in the presence of water indicated the nonconducive nature of gelation in very highly polar solvents, methanol, and DMF or, in very low polarity solvents, such as, cyclohexane and carbon tetrachloride.
Sigma-Aldrich
6-Aminocaproic acid, SAJ special grade, ≥99.0%
Supelco
Aminocaproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
6-Aminocaproic acid, BioUltra, ≥99%
Sigma-Aldrich
6-Aminocaproic acid, ≥99% (titration), powder
Sigma-Aldrich
6-Aminohexanoic acid, ≥98.5% (NT)