Merck
  • Positron emission tomography of human hepatocellular carcinoma xenografts in mice using copper (II)-64 chloride as a tracer with copper (II)-64 chloride.
Positron emission tomography of human hepatocellular carcinoma xenografts in mice using copper (II)-64 chloride as a tracer with copper (II)-64 chloride.
Academic radiology (2011-11-08)
Haiyuan Zhang, Huawei Cai, Xin Lu, Otto Muzik, Fangyu Peng
ABSTRACT
The aim of this study was to assess copper metabolism of human hepatocellular carcinoma (HCC) with positron emission tomographic (PET) imaging using copper (II)-64 chloride ((64)CuCl(2)) as a tracer. PET imaging of athymic mice (n = 5) bearing extrahepatic HCC xenografts was performed 24 hours after the intravenous injection of (64)CuCl(2), followed by ex vivo tissue radioactivity assay. Expression of human copper transporter 1 (hCTR1) in HCC cells and tissues was examined by real-time reverse transcription polymerase chain reaction and immunohistochemistry analysis, respectively. The extrahepatic HCC xenografts in mice with increased uptake of (64)Cu radionuclide were visualized on the micro-PET images obtained 24 hours after the intravenous injection of (64)CuCl(2). PET quantitative analysis revealed increased (64)Cu radioactivity in tumor tissues (2.7 ± 0.6 %ID/g) compared to that in the soft tissue of the left shoulder opposite to the tumor site (0.6 ± 0.2 %ID/g) and the brain (0.7 ± 0.1 %ID/g) but lower than that of the liver (16.6 ± 1.3 %ID/g). Expression of hCTR1 in the HCC cells and xenograft tumor tissues was demonstrated by real-time reverse transcription polymerase chain reaction and immunohistochemistry analysis, respectively. The expression level of hCTR1 in the Hep3B HCC xenograft tissues was lower than that detected in the normal hepatic tissues and the tissue samples of well-differentiated primary HCC. Variable expression of hCTR1 was detected in the tissue samples of moderately differentiated primary HCC. Extrahepatic human HCC xenografts in mice could be localized with (64)CuCl(2) PET imaging, which might be useful for the localization and quantitative assessment of copper metabolism in extrahepatic metastases of HCC in humans.
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads, ≥99.99% trace metals basis
Sigma-Aldrich
Copper(I) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(I) chloride, JIS special grade, ≥95.0%
Sigma-Aldrich
Copper(I) chloride, ReagentPlus®, purified, ≥99%
Sigma-Aldrich
Copper(I) chloride, reagent grade, 97%