Merck
  • Supramolecular chemical sensors based on pyrene monomer-excimer dual luminescence.
Supramolecular chemical sensors based on pyrene monomer-excimer dual luminescence.
Chemistry, an Asian journal (2011-01-29)
Sekar Karuppannan, Jean-Claude Chambron
ABSTRACT
The past ten years have seen a spectacular development of chemical sensors based on the monomer-excimer dual luminescence of aromatic systems, such as pyrene. Either in the form of integrated or multicomponent molecular devices these chemosensors have been attracting a high interest above all because of their unique ratiometric properties. This review will focus on the latter systems, which can be classified into two classes: Firstly, the assembly of receptor-effector conjugates is triggerred by the analyte of interest. As a result, the sensor shows monomer to excimer fluorescence switching upon substrate binding. Secondly, the supramolecular assembly that constitutes the sensor is perturbed by interaction with the analyte. This induces a conformational change or the exchange of a component of the system, which is the cause of the luminescence switch effect.
Sigma-Aldrich
γ-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, Wacker Chemie AG, ≥90.0% cyclodextrin basis (HPLC)