Merck
  • [Prenatal sex identification with biotinylated human Y-specific DNA probe].
[Prenatal sex identification with biotinylated human Y-specific DNA probe].
Hua xi yi ke da xue xue bao = Journal of West China University of Medical Sciences = Huaxi yike daxue xuebao (1990-09-01)
W Dong, S Jiang, S Zhang, L Zhao
ABSTRACT
Using biotinylated human Y-specific DNA probe PHY2.1 and Cos 84, we carried out Southern and dot hybridization with human DNA from artificially aborted chorionic villi, and with DNA of peripheral lymphocytes from normal male and female. The results show that PHY2.1 may be applied in prenatal sex identification both by Southern and by dot hybridization but with Cos 84, only Southern hybridization could be used.
Sigma-Aldrich
Sodium p-toluenesulfinate, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium p-toluenesulfinate, purum, anhydrous, ≥96.0% (NT)
Pricing and availability is not currently available.