Merck
A high-throughput screen for alpha particle radiation protectants.
Assay and drug development technologies (2010-07-28)
Jonathan H Seideman, David Shum, Hakim Djaballah, David A Scheinberg
ABSTRACT
Alpha-particle-emitting elements are of increasing importance as environmental and occupational carcinogens, toxic components of radiation dispersal devices and accidents, and potent therapeutics in oncology. Alpha particle radiation differs from radiations of lower linear energy transfer in that it predominantly damages DNA via direct action. Because of this, radical scavengers effective for other radiations have had only limited effect in mitigating alpha particle toxicity. We describe here a simple assay and a pilot screen of 3,119 compounds in a high-throughput screen (HTS), using the alpha-particle-emitting isotope, ²²⁵Ac, for the discovery of compounds that might protect mammalian cells from alpha particles through novel mechanisms. The assay, which monitored the viability of a myeloid leukemic cell line upon alpha particle exposure, was robust and reproducible, yielding a Z' factor of 0.66 and a signal-to-noise ratio of nearly 10 to 1. Surprisingly, 1 compound emerged from this screen, epoxy-4,5-α-dihydroxysantonin (EDHS), that showed considerable protective activity. While the value of EDHS remains to be determined, its discovery is a proof of concept and validation of the utility of this HTS methodology. Further application of the described assay could yield compounds useful in minimizing the toxicity and carcinogenesis associated with alpha particle exposure.
Sigma-Aldrich
Aluminum nitrate nonahydrate, 99.997% trace metals basis
Sigma-Aldrich
Aluminum nitrate nonahydrate, JIS special grade, ≥98.0%
Sigma-Aldrich
Aluminum nitrate nonahydrate, ACS reagent, ≥98%