Merck
  • Determination of the metabolites of the new designer drugs bk-MBDB and bk-MDEA in human urine.
Determination of the metabolites of the new designer drugs bk-MBDB and bk-MDEA in human urine.
Forensic science international (2009-05-02)
Kei Zaitsu, Munehiro Katagi, Hiroe T Kamata, Tooru Kamata, Noriaki Shima, Akihiro Miki, Hitoshi Tsuchihashi, Yasushige Mori
ABSTRACT
This is the first report on identifying the specific metabolites of the new designer drugs 2-methylamino-1-(3,4-methylenedioxyphenyl)butan-1-one (bk-MBDB) and 2-ethylamino-1-(3,4-methylenedioxyphenyl)propan-1-one (bk-MDEA) in human urine using synthesized standards. Based on GC/MS and LC/MS, we identified N-dealkylation, demethylenation followed by O-methylation, and beta-ketone reduction as their major metabolic pathways. The quantitative analyses by LC/MS revealed that both demethylenation followed by O-methylation and beta-ketone reduction were superior to N-dealkylation and that both bk-MBDB and bk-MDEA were mainly metabolized into their corresponding 4-hydroxy-3-methoxy metabolites (4-OH-3MeO metabolites). After hydrolysis, the concentrations of 4-OH-3MeO metabolites and 3-hydroxy-4-methoxy metabolites of both bk-MBDB and bk-MDEA dramatically increased, suggesting that the metabolites mainly exist as their conjugates.
Product Description
Supelco
(±)-MDEA solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(±)-3,4-Methylenedioxy-N-­ethyl­amphetamine hydrochloride, ≥98% (TLC)