Merck
  • (+)-Pinoresinol is a putative hypoglycemic agent in defatted sesame (Sesamum indicum) seeds though inhibiting α-glucosidase.
(+)-Pinoresinol is a putative hypoglycemic agent in defatted sesame (Sesamum indicum) seeds though inhibiting α-glucosidase.
Bioorganic & medicinal chemistry letters (2012-07-24)
Arin Wikul, Thanakorn Damsud, Keita Kataoka, Preecha Phuwapraisirisan
ABSTRACT
Defatted sesame seeds have been reported for hypoglycemic effect in mice and T2DM women. An attempted to identify active components responsible for this effect was conducted using α-glucosidase-guided fractionation, resulting in the isolation of various lignans. Of compounds isolated, only (+)-pinoresinol showed inhibitory activity against rat intestinal maltase with an IC(50) value of 34.3 μM. The kinetic study indicated that enzymatic hydrolysis of maltose is inhibited by (+)-pinoresinol through competitive and noncompetitive manners. However, a lower dissociation constant (k(i) 288 M) of EI complex suggested that competitive inhibition is predominant over noncompetitive mode (k'(i) 1342 M).
Product Description
Supelco
Pinoresinol, analytical standard
Sigma-Aldrich
Pinoresinol, ≥95.0% (HPLC)