Merck
  • Deciphering lineage specification during early embryogenesis in mouse gastruloids using multilayered proteomics.
Deciphering lineage specification during early embryogenesis in mouse gastruloids using multilayered proteomics.
Cell stem cell (2024-05-17)
Suzan Stelloo, Maria Teresa Alejo-Vinogradova, Charlotte A G H van Gelder, Dick W Zijlmans, Marek J van Oostrom, Juan Manuel Valverde, Lieke A Lamers, Teja Rus, Paula Sobrevals Alcaraz, Tilman Schäfers, Cristina Furlan, Pascal W T C Jansen, Marijke P A Baltissen, Katharina F Sonnen, Boudewijn Burgering, Maarten A F M Altelaar, Harmjan R Vos, Michiel Vermeulen
ABSTRACT
Gastrulation is a critical stage in embryonic development during which the germ layers are established. Advances in sequencing technologies led to the identification of gene regulatory programs that control the emergence of the germ layers and their derivatives. However, proteome-based studies of early mammalian development are scarce. To overcome this, we utilized gastruloids and a multilayered mass spectrometry-based proteomics approach to investigate the global dynamics of (phospho) protein expression during gastruloid differentiation. Our findings revealed many proteins with temporal expression and unique expression profiles for each germ layer, which we also validated using single-cell proteomics technology. Additionally, we profiled enhancer interaction landscapes using P300 proximity labeling, which revealed numerous gastruloid-specific transcription factors and chromatin remodelers. Subsequent degron-based perturbations combined with single-cell RNA sequencing (scRNA-seq) identified a critical role for ZEB2 in mouse and human somitogenesis. Overall, this study provides a rich resource for developmental and synthetic biology communities endeavoring to understand mammalian embryogenesis.
Product Description
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Anti-HOXC8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution