Merck
  • RNA polymerase II dynamics and mRNA stability feedback scale mRNA amounts with cell size.
RNA polymerase II dynamics and mRNA stability feedback scale mRNA amounts with cell size.
Cell (2023-11-10)
Matthew P Swaffer, Georgi K Marinov, Huan Zheng, Lucas Fuentes Valenzuela, Crystal Yee Tsui, Andrew W Jones, Jessica Greenwood, Anshul Kundaje, William J Greenleaf, Rodrigo Reyes-Lamothe, Jan M Skotheim
ABSTRACT
A fundamental feature of cellular growth is that total protein and RNA amounts increase with cell size to keep concentrations approximately constant. A key component of this is that global transcription rates increase in larger cells. Here, we identify RNA polymerase II (RNAPII) as the limiting factor scaling mRNA transcription with cell size in budding yeast, as transcription is highly sensitive to the dosage of RNAPII but not to other components of the transcriptional machinery. Our experiments support a dynamic equilibrium model where global RNAPII transcription at a given size is set by the mass action recruitment kinetics of unengaged nucleoplasmic RNAPII to the genome. However, this only drives a sub-linear increase in transcription with size, which is then partially compensated for by a decrease in mRNA decay rates as cells enlarge. Thus, limiting RNAPII and feedback on mRNA stability work in concert to scale mRNA amounts with cell size.
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-5) Antibody, clone 3E8, culture supernatant, clone 3E8, from rat
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys27) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-RNA Polymerase II Antibody, CTD Antibody, clone 8WG16, clone 8WG16, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-acetyl-Histone H4 (Lys16) Antibody, Upstate®, from rabbit