Merck
  • Understanding cell fate acquisition in stem-cell-derived pancreatic islets using single-cell multiome-inferred regulomes.
Understanding cell fate acquisition in stem-cell-derived pancreatic islets using single-cell multiome-inferred regulomes.
Developmental cell (2023-04-12)
Han Zhu, Gaowei Wang, Kim-Vy Nguyen-Ngoc, Dongsu Kim, Michael Miller, Georgina Goss, Jenna Kovsky, Austin R Harrington, Diane C Saunders, Alexander L Hopkirk, Rebecca Melton, Alvin C Powers, Sebastian Preissl, Francesca M Spagnoli, Kyle J Gaulton, Maike Sander
ABSTRACT
Pancreatic islet cells derived from human pluripotent stem cells hold great promise for modeling and treating diabetes. Differences between stem-cell-derived and primary islets remain, but molecular insights to inform improvements are limited. Here, we acquire single-cell transcriptomes and accessible chromatin profiles during in vitro islet differentiation and pancreas from childhood and adult donors for comparison. We delineate major cell types, define their regulomes, and describe spatiotemporal gene regulatory relationships between transcription factors. CDX2 emerged as a regulator of enterochromaffin-like cells, which we show resemble a transient, previously unrecognized, serotonin-producing pre-β cell population in fetal pancreas, arguing against a proposed non-pancreatic origin. Furthermore, we observe insufficient activation of signal-dependent transcriptional programs during in vitro β cell maturation and identify sex hormones as drivers of β cell proliferation in childhood. Altogether, our analysis provides a comprehensive understanding of cell fate acquisition in stem-cell-derived islets and a framework for manipulating cell identities and maturity.
Sigma-Aldrich
γ-Secretase Inhibitor XX, This γ-secretase inhibitor, CAS 209984-56-5, is a cell-permeable dibenzazepine compound that lowers both brain and plasma Aβ40 levels by ~72% in Tg2576 mutant APP transgenic mouse model.
Sigma-Aldrich
Anti-SLC18A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
α-Amyloid Precursor Protein Modulator, A cell-permeable benzolactam derived PKC activator (Ki = 11.9 nM for PKCα) that efficiently enhances non-amyloidogenic α-processing of amyloid precursor protein (APP) even at 100 nM in human fibroblast AG06848.
Sigma-Aldrich
Anti-Tryptophan Hydroxylase Antibody, Chemicon®, from sheep
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
Sigma-Aldrich
SANT-1, ≥98% (HPLC), powder