Merck
  • Runx2 phosphorylation induced by fibroblast growth factor-2/protein kinase C pathways.
Runx2 phosphorylation induced by fibroblast growth factor-2/protein kinase C pathways.
Proteomics (2006-01-20)
Byung-Gyu Kim, Hyun-Jung Kim, Hye-Jeong Park, Youn-Jeong Kim, Won-Joon Yoon, Seung-Jin Lee, Hyun-Mo Ryoo, Je-Yoel Cho
ABSTRACT
Runx2 is a key transcription factor in osteoblast differentiation, and its activity is regulated by fibroblast growth factors (FGFs). Craniosynostosis, characterized by premature suture closure, results from mutations that generate constitutively active FGF receptors (FGFRs). We previously showed that FGF/FGFR-activated protein kinase C (PKC) is involved in the expression and activity of Runx2. Activated PKCdelta physically interacts with Runx2 in FGF2-stimulated MC3T3-E1 preosteoblastic cells. Immunopurified Runx2 protein reacted with PKCdelta kinase, and a phosphorylated 1460-Da peptide fragment (amino acids 241-252, 1380-Da) derived from Runx2 was also detected in MS analysis. Computer analysis predicted that Ser247 in this Runx2 can be a possible phosphorylation site by PKCdelta. We also showed that Runx2 activity after FGF stimulation correlates with the presence of the Runx2 Ser247 residue. The S247A (Ser --> Ala) mutation confers decreased transcriptional activity on a Runx2-responsive promoter after FGF treatment.
Product Description
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Protein LoBind Microplate 384/V-PP, PCR clean, V-bottom clear wells, white border, pack of 240 ea (10 bags × 24 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 384 wells, white plate, conical bottom, colorless wells, capacity 200 μL, pack of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 80 ea (10 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 384 wells, white plate, conical bottom, colorless wells, capacity 200 μL, pack of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® DNA LoBind tubes, capacity 1.5 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® DNA LoBind tubes, capacity 0.5 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® DNA LoBind tubes, capacity 2.0 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 80 ea (10 bags × 8 plates)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, yellow plate, conical bottom, colorless wells, capacity 500 μL, pkg of 20 ea (5 bags × 4 plates)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, yellow plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Protein LoBind tubes, capacity 0.5 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Protein LoBind tubes, capacity 1.5 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Protein LoBind tubes, capacity 2.0 mL, PCR clean, pkg of 100 ea (2 x 50ea)