Merck
The prophase oocyte nucleus is a homeostatic G-actin buffer.
Journal of cell science (2022-02-04)
Kathleen Scheffler, Federica Giannini, Tom Lemonnier, Binyam Mogessie
ABSTRACT
Formation of healthy mammalian eggs from oocytes requires specialised F-actin structures. F-actin disruption produces aneuploid eggs, which are a leading cause of human embryo deaths, genetic disorders and infertility. We found that oocytes contain prominent nuclear F-actin structures that are correlated with meiotic developmental capacity. We demonstrate that nuclear F-actin is a conserved feature of healthy mammalian oocytes and declines significantly with female reproductive ageing. Actin monomers used for nuclear F-actin assembly are sourced from an excess pool in the oocyte cytoplasm. Increasing monomeric G-actin transfer from the cytoplasm to the nucleus or directly enriching the nucleus with monomers led to assembly of stable nuclear F-actin bundles that significantly restrict chromatin mobility. By contrast, reducing G-actin monomer transfer by blocking nuclear import triggered assembly of a dense cytoplasmic F-actin network that is incompatible with healthy oocyte development. Overall, our data suggest that the large oocyte nucleus helps to maintain cytoplasmic F-actin organisation and that defects in this function are linked with reproductive age-related female infertility. This article has an associated First Person interview with Federica Giannini, joint first author of the paper.
Sigma-Aldrich
Arp2/3 Complex Inhibitor I, CK-666, Arp2/3 Complex Inhibitor I, CK-666, CAS 442633-00-3, is a cell-permeable selective inhibitor of actin assembly mediated by actin-related protein Arp2/3 complex (IC50 = 4 uM in human).
Sigma-Aldrich
dTAG-13, ≥98% (HPLC)
Sigma-Aldrich
Latrunculin B, Latrunculia magnifica, Latrunculin B, CAS 76343-94-7, is a unique marine toxin that inhibits actin polymerization and disrupts microfilament organization. It is 10 to 100-fold more potent than cytochalasins.