Merck
  • Low Expression of Estrogen Receptor-α and Progesterone Receptor in Human Breast Cancer Tissues Is Associated With High-Grade Human Cytomegalovirus Protein Expression.
Low Expression of Estrogen Receptor-α and Progesterone Receptor in Human Breast Cancer Tissues Is Associated With High-Grade Human Cytomegalovirus Protein Expression.
Clinical breast cancer (2017-06-10)
Afsar Rahbar, Joel Touma, Helena Costa, Belghis Davoudi, Ida Rashid Bukholm, Torill Sauer, Katja Vetvik, Jürgen Geisler, Cecilia Söderberg-Naucler
ABSTRACT
The underlying mechanisms for breast cancer (BC) are largely unknown. We investigated possible correlations between the expression levels of human cytomegalovirus (HCMV) proteins and established histopathological markers of BC, including expression of estrogen receptor (ER)-α, the progesterone receptor (PgR), and HER2. We retrospectively examined paraffin-embedded biopsy specimens of BC (n = 62), ductal carcinoma in situ (n = 19), and adjacent normal breast tissue (n = 42) for HCMV immediate-early protein (IE), HCMV late antigen, HCMV DNA and RNA, and investigated possible correlations between them and expression of ER-α, PgR, and HER2. HCMV DNA and RNA were detected in all examined infiltrating BCs. High-grade positivity for HCMV-IE was detected in 77% of infiltrating BCs, 39% of ductal carcinomas in situ, and 7% of tumor-free breast tissue samples. HCMV expression correlated inversely with ER-α (P = .02) and PgR (P = .003) expression. HER2 expression was also reduced in HCMV-positive samples without reaching a level of statistical significance (P = .09). The negative correlation between high-grade expression HCMV-IE and hormone receptor expression suggests a role for HCMV in hormone receptor-negative BC tumors, possibly by forcing BC cells into a more aggressive phenotype.
Sigma-Aldrich
Anti-Cytomegalovirus Antibody, late Antigen, clone 1G5.2, clone 1G5.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Cytomegalovirus Antibody, clone 8B1.2, clone 8B1.2, Chemicon®, from mouse