Merck
  • Design and evaluation of anacardic acid derivatives as anticavity agents.
Design and evaluation of anacardic acid derivatives as anticavity agents.
European journal of medicinal chemistry (2007-10-26)
Ivan R Green, Felismino E Tocoli, Sang Hwa Lee, Ken-ichi Nihei, Isao Kubo
ABSTRACT
On the basis of antibacterial anacardic acids, 6-pentadecenylsalicylic acids, isolated from the cashew apple, Anacardium occidentale L. (Anacardiaceae), a series of 6-alk(en)ylsalicylic acids were synthesized and tested for their antibacterial activity against Streptococcus mutans ATCC 25175. Among them, 6-(4',8'-dimethylnonyl)salicylic acid was found to exhibit the most potent antibacterial activity against this cariogenic bacterium with the minimum inhibition concentration (MIC) of 0.78 microg/ml.
Product Description
Supelco
Ginkgolic acid C15:1, analytical standard
Supelco
trans-Anethole, analytical standard
Sigma-Aldrich
trans-Anethole, 99%
Sigma-Aldrich
trans-Anethole, ≥99%, FCC, FG