Merck
Effect of SOX2 Repression on Corneal Endothelial Cells.
International journal of molecular sciences (2020-06-25)
Jin Sun Hwang, Ho Chul Yi, Young Joo Shin
ABSTRACT
Human corneal endothelial cells (hCECs) pump out water from the stroma and maintain the clarity of the cornea. The sex-determining region Y-box 2 (SOX2) participates in differentiation during the development of the anterior segment of the eye and is found in the periphery of wounded corneas. This study was performed to investigate the effect of SOX2 repression on hCECs. Cultured hCECs were transfected by siRNA for SOX2. The wound healing rate and cell viability were measured. The cell proliferation-associated protein level was evaluated by Western blotting and RT-PCR. The energy production and mitochondrial function were measured, and cell shape and WNT signaling were assessed. Upon transfecting the cultured cells with siRNA for SOX2, the SOX2 level was reduced by 80%. The wound healing rate and viability were also reduced. Additionally, CDK1, cyclin D1, SIRT1, and ATP5B levels were reduced, and CDKN2A and pAMPK levels were increased. Mitochondrial oxidative stress and mitochondrial viability decreased, and the cell shape became elongated. Furthermore, SMAD1, SNAI1, WNT3A, and β-catenin levels were increased. SOX2 repression disrupts the normal metabolism of hCECs through modulating WNT signaling and mitochondrial functions.
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Anti-INK4a (p16) Antibody, clone 13H4.1, clone 13H4.1, from mouse