Merck
  • A versatile depigmentation, clearing, and labeling method for exploring nervous system diversity.
A versatile depigmentation, clearing, and labeling method for exploring nervous system diversity.
Science advances (2020-06-12)
Marko Pende, Karim Vadiwala, Hannah Schmidbaur, Alexander W Stockinger, Prayag Murawala, Saiedeh Saghafi, Marcus P S Dekens, Klaus Becker, Roger Revilla-I-Domingo, Sofia-Christina Papadopoulos, Martin Zurl, Pawel Pasierbek, Oleg Simakov, Elly M Tanaka, Florian Raible, Hans-Ulrich Dodt
ABSTRACT
Tissue clearing combined with deep imaging has emerged as a powerful alternative to classical histological techniques. Whereas current techniques have been optimized for imaging selected nonpigmented organs such as the mammalian brain, natural pigmentation remains challenging for most other biological specimens of larger volume. We have developed a fast DEpigmEntation-Plus-Clearing method (DEEP-Clear) that is easily incorporated in existing workflows and combines whole system labeling with a spectrum of detection techniques, ranging from immunohistochemistry to RNA in situ hybridization, labeling of proliferative cells (EdU labeling) and visualization of transgenic markers. With light-sheet imaging of whole animals and detailed confocal studies on pigmented organs, we provide unprecedented insight into eyes, whole nervous systems, and subcellular structures in animal models ranging from worms and squids to axolotls and zebrafish. DEEP-Clear thus paves the way for the exploration of species-rich clades and developmental stages that are largely inaccessible by regular imaging approaches.
Product Description
Supelco
Antipyrine, analytical standard
Sigma-Aldrich
N,N,N′,N′-Tetrakis(2-hydroxyethyl)ethylenediamine, technical
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
Meglumine diatrizoate, organic