Merck
Gastrin: an acid-releasing, proliferative and immunomodulatory peptide?
Mini reviews in medicinal chemistry (2010-04-13)
Sara Calatayud, Angeles Alvarez, Víctor M Víctor
ABSTRACT
Gastrin release is affected by gastric inflammatory conditions. Antral G cells respond to inflammatory mediators by increasing gastrin secretion. Accumulating experimental evidence suggests that gastrin exerts immunomodulatory and proinflammatory effects. Gastrin could be a contributing factor to these pathologies, which may constitute a new justification for pharmacological blockade of gastrin action.
Sigma-Aldrich
[Gly18]-Gastrin 1-17 human
Sigma-Aldrich
Gastrin I, Mini human
Sigma-Aldrich
Gastrin I (1-14), ≥92% (HPLC)
Sigma-Aldrich
[Leu15,Gly18]-Gastrin2-17