Merck
  • Vascular smooth muscle cells ablation with endovascular nonthermal irreversible electroporation.
Vascular smooth muscle cells ablation with endovascular nonthermal irreversible electroporation.
Journal of vascular and interventional radiology : JVIR (2010-10-12)
Elad Maor, Antoni Ivorra, James J Mitchell, Boris Rubinsky
ABSTRACT
To evaluate the effect of endovascular nonthermal irreversible electroporation (NTIRE) on blood vessels. Specially made endovascular devices with four electrodes on top of inflatable balloons were used to apply electroporation pulses. Finite element simulations were used to characterize NTIRE protocols that would not induce thermal damage to treated tissues. Right iliac arteries of eight rabbits were treated with 90 NTIRE pulses. Angiograms were performed before and after the procedures. Arterial specimens were harvested at 7 and 35 days. Evaluation included hematoxylin and eosin, elastic von Giessen, and Masson trichrome stains. Immunohistochemistry of selected slides included smooth muscle actin (SMA), proliferating cell nuclear antigen, von Willebrand factor (VWF), and S-100 antigen. At 7 days, all NTIRE-treated arterial segments displayed complete, transmural ablation of vascular smooth muscle cells (VSMC). At 35 days, similar damage to VSMC was noted. In most cases, the elastic lamina remained intact, and endothelial layer regenerated. Occasional mural inflammation and cartilaginous metaplasia were noted. After 5 weeks, there was no evidence of significant VSMC proliferation, with the dominant process being wall fibrosis with regenerated endothelium. NTIRE can be applied in an endovascular approach. It efficiently ablates vessel wall within seconds and with no damage to extracellular structures. NTIRE has possible applications in many fields of clinical cardiology, including arterial restenosis and cardiac arrhythmias.
Sigma-Aldrich
Anti-S-100 Protein Antibody, clone 15E2E2, clone 15E2E2, Chemicon®, from mouse