Merck
  • Novel vitamin E forms in leaves of Kalanchoe daigremontiana and Phaseolus coccineus.
Novel vitamin E forms in leaves of Kalanchoe daigremontiana and Phaseolus coccineus.
Journal of plant physiology (2011-08-23)
Jerzy Kruk, Adam Pisarski, Renata Szymańska
ABSTRACT
In the present study, we isolated novel tocochromanols from green leaves of Kalanchoe daigremontiana and primary leaves of etiolated seedlings of Phaseolus coccineus that were identified as β-, γ-, and δ-tocomonoenols with unsaturation at the terminal isoprene unit of the side chain. The content of γ-tocomonoenol in leaves of etiolated bean increased gradually with the age of seedlings, reaching 50% of the γ-tocopherol level in 40-day-old plants. The content of this compound in leaves was increased by short illumination of etiolated plants and by addition of homogentisic acid, a biosynthetic precursor of tocopherols. These data indicated that γ-tocomonoenol is synthesized de novo from homogentisic acid and tetrahydro-geranylgeraniol diphosphate, a phytol precursor. Based on these results, a biosynthetic pathway of tocomonoenols is proposed.
Sigma-Aldrich
(+)-γ-Tocopherol, ≥96% (HPLC)
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g