Merck
  • RpA1 ameliorates symptoms of mutant ataxin-1 knock-in mice and enhances DNA damage repair.
RpA1 ameliorates symptoms of mutant ataxin-1 knock-in mice and enhances DNA damage repair.
Human molecular genetics (2017-02-09)
Juliana Bosso Taniguchi, Kanoh Kondo, Kyota Fujita, Xigui Chen, Hidenori Homma, Takeaki Sudo, Ying Mao, Kei Watase, Toshihiro Tanaka, Kazuhiko Tagawa, Takuya Tamura, Shin-Ichi Muramatsu, Hitoshi Okazawa
ABSTRACT
DNA damage and repair is a critical domain of many neurodegenerative diseases. In this study, we focused on RpA1, a candidate key molecule in polyQ disease pathologies, and tested the therapeutic effect of adeno-associated virus (AAV) vector expressing RpA1 on mutant Ataxin-1 knock-in (Atxn1-KI) mice. We found significant effects on motor functions, normalized DNA damage markers (γH2AX and 53BP1), and improved Purkinje cell morphology; effects that lasted for 50 weeks following AAV-RpA1 infection. In addition, we confirmed that AAV-RpA1 indirectly recovered multiple cellular functions such as RNA splicing, transcription and cell cycle as well as abnormal morphology of dendrite and dendritic spine of Purkinje cells in Atxn1-KI mice. All these results suggested a possibility of gene therapy with RpA1 for SCA1.
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Calbindin-D-28K (EG-20) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Ataxin-1 Antibody, 11NQ, clone N76/8, clone N76/8, from mouse
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse