Merck
  • GREB1 induced by Wnt signaling promotes development of hepatoblastoma by suppressing TGFβ signaling.
GREB1 induced by Wnt signaling promotes development of hepatoblastoma by suppressing TGFβ signaling.
Nature communications (2019-08-30)
Shinji Matsumoto, Taku Yamamichi, Koei Shinzawa, Yuuya Kasahara, Satoshi Nojima, Takahiro Kodama, Satoshi Obika, Tetsuo Takehara, Eiichi Morii, Hiroomi Okuyama, Akira Kikuchi
ABSTRACT
The β-catenin mutation is frequently observed in hepatoblastoma (HB), but the underlying mechanism by which Wnt/β-catenin signaling induces HB tumor formation is unknown. Here we show that expression of growth regulation by estrogen in breast cancer 1 (GREB1) depends on Wnt/β-catenin signaling in HB patients. GREB1 is localized to the nucleus where it binds Smad2/3 in a competitive manner with p300 and inhibits TGFβ signaling, thereby promoting HepG2 HB cell proliferation. Forced expression of β-catenin, YAP, and c-Met induces HB-like mouse liver tumor (BYM mice), with an increase in GREB1 expression and HB markers. Depletion of GREB1 strongly suppresses marker gene expression and HB-like liver tumorigenesis, and instead enhances TGFβ signaling in BYM mice. Furthermore, antisense oligonucleotides for GREB1 suppress the formation of HepG2 cell-induced tumors and HB-like tumors in vivo. We propose that GREB1 is a target molecule of Wnt/β-catenin signaling and required for HB progression.
Sigma-Aldrich
Anti-phospho-Catenin-β (pTyr654) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GREB1 Antibody, clone GREB1Ab, clone GREB1Ab, from mouse
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-TCF-4 Antibody, clone 6H5-3, clone 6H5-3, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit