Merck
  • Molecular basis of senescence transmitting in the population of human endometrial stromal cells.
Molecular basis of senescence transmitting in the population of human endometrial stromal cells.
Aging (2019-11-07)
Anastasiia Griukova, Pavel Deryabin, Alla Shatrova, Elena Burova, Valeria Severino, Annarita Farina, Nikolay Nikolsky, Aleksandra Borodkina
ABSTRACT
Hormone-regulated proliferation and differentiation of endometrial stromal cells (ESCs) determine overall endometrial plasticity and receptivity to embryos. Previously we revealed that ESCs may undergo premature senescence, accompanied by proliferation loss and various intracellular alterations. Here we focused on whether and how senescence may be transmitted within the ESCs population. We revealed that senescent ESCs may induce paracrine senescence in young counterparts via cell contacts, secreted factors and extracellular vesicles. According to secretome-wide profiling we identified plasminogen activator inhibitor -1 (PAI-1) to be the most prominent protein secreted by senescent ESCs (data are available via ProteomeXchange with identifier PXD015742). By applying CRISPR/Cas9 techniques we disclosed that PAI-1 secreted by senescent ESCs may serve as the master-regulator of paracrine senescence progression within the ESCs population. Unraveled molecular basis of senescence transduction in the ESCs population may be further considered in terms of altered endometrial plasticity and sensitivity to invading embryo, thus contributing to the female infertility curing.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-Hsp70/Hsp72 Antibody, clone 2H9, clone 2H9, from mouse
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse