Merck
Axonal Odorant Receptors Mediate Axon Targeting.
Cell reports (2019-12-26)
Ilaria Zamparo, Simona Francia, Sira Angela Franchi, Nelly Redolfi, Elisa Costanzi, Axelle Kerstens, Yosuke Fukutani, Roberto Battistutta, Patrizia Polverino de Laureto, Sebastian Munck, Bart De Strooper, Hiroaki Matsunami, Claudia Lodovichi
ABSTRACT
In mammals, odorant receptors not only detect odors but also define the target in the olfactory bulb, where sensory neurons project to give rise to the sensory map. The odorant receptor is expressed at the cilia, where it binds odorants, and at the axon terminal. The mechanism of activation and function of the odorant receptor at the axon terminal is, however, still unknown. Here, we identify phosphatidylethanolamine-binding protein 1 as a putative ligand that activates the odorant receptor at the axon terminal and affects the turning behavior of sensory axons. Genetic ablation of phosphatidylethanolamine-binding protein 1 in mice results in a strongly disturbed olfactory sensory map. Our data suggest that the odorant receptor at the axon terminal of olfactory neurons acts as an axon guidance cue that responds to molecules originating in the olfactory bulb. The dual function of the odorant receptor links specificity of odor perception and axon targeting.
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Supelco
(±)-Sulfinpyrazone, analytical standard
Sigma-Aldrich
2-Coumaranone, 97%
Sigma-Aldrich
(S)-(+)-Carvone, 96%
Sigma-Aldrich
2-Isobutyl-3-methoxypyrazine, 99%
Sigma-Aldrich
Menthone, mixture of isomers, ≥97.0% (GC)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Supelco
Menthone, analytical standard